Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation
70.75€ - 172.00€
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11411078
|
Grant Instruments™
PRS-10 |
10 Tubes up to 30 mm, Tube volumes up to 50 mL |
71.75€
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
11421028
|
Grant Instruments™
PRS-14 |
14 tubes up to 30mm, tube volumes up to 50mL |
128.00€
Each |
In Stock
Log in to see stock. |
|||||
11431028
|
Grant Instruments™
PRS-22 |
22 Tubes up to 15 mm, Tube volumes up to 15 mL |
96.35€
Each |
In Stock
Log in to see stock. |
|||||
11491068
|
Grant Instruments™
PRS-26 |
26 Tubes up to 15 mm, 1.5 -15 mL |
107.00€
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
11411028
|
Grant Instruments™
PRS-48 |
48 Tubes up to Ø 15 mm, Tube volumes 1.5-15 mL |
172.00€
Each |
In Stock
Log in to see stock. |
Hai già un account? Accedi per visualizzare il prezzo e la disponibilità. Sei nuovo da queste parti? Registrati qui! |
||||
10860891
|
Grant Instruments™
PRS-5-12 |
5 Tubes up to 30 mm and 12 Tubes up to 15 mm, Tube volumes 50 mL/1.5-15 mL |
70.75€
Each |
In Stock
Log in to see stock. |
Hai già un account? Accedi per visualizzare il prezzo e la disponibilità. Sei nuovo da queste parti? Registrati qui! |
||||
12915511
|
Grant Instruments™
PRS-8-22 |
8 tubes up to 30 mm plus 22 tubes up to 15 mm, tube volumes up to 50 mL/1.5-15 mL |
128.00€
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
Hai già un account? Accedi per visualizzare il prezzo e la disponibilità. Sei nuovo da queste parti? Registrati qui! |
||||
Descrizione
- For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
- Hold as few as 8 test tubes or as many as 48.
- Accommodates a variety of capacities
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.