Learn More
Buerkle™ Plastic Griffin Beakers with Blue Graduations
Manufactured from highly transparent polypropylene.
20.90 SEK - 370.35 SEK
Specifications
Color | Translucent |
---|---|
For Use With (Application) | Various Uses in The Laboratory |
Type | Griffin Beaker |
Autoclavable | Autoclavable |
Graduated | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10159261
|
Buerkle™
7122-0050 |
50 mL |
40.30 SEK
Each |
Estimated Shipment: 10-07-2025
Log in to see stock. |
|||||
17280674
|
Buerkle™
7122-0025 |
25 mL |
20.90 SEK
Each |
In Stock
Log in to see stock. |
|||||
10183515
|
Buerkle™
7122-0150 |
150 mL |
40.30 SEK
Each |
In Stock
Log in to see stock. |
|||||
10435572
|
Buerkle™
7122-0500 |
500 mL |
50.28 SEK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
|||||
10750773
|
Buerkle™
7122-0600 |
600 mL |
52.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
10589502
|
Buerkle™
7122-1000 |
1000 mL |
78.68 SEK
Each |
In Stock
Log in to see stock. |
|||||
10287427
|
Buerkle™
7122-3000 |
3000 mL |
285.92 SEK
Each |
In Stock
Log in to see stock. |
|||||
10462888
|
Buerkle™
7122-5000 |
5000 mL |
370.35 SEK
Each |
In Stock
Log in to see stock. |
|||||
Description
- For various uses
- Acc. to DIN 12331 and ISO 7056
- Blue graduation printed on
- Shatterproof
- Autoclavable
Specifications
Translucent | |
Griffin Beaker | |
Yes |
Various Uses in The Laboratory | |
Autoclavable |