Learn More
Fisherbrand™ Single-Channel Pipette
Aspirates and dispenses viscous liquids accurately and reliably. Fisherbrand™ Single-Channel Pipette include a lock counter to prevent accidental volume changes. Lightweight, ergonomic design minimizes repetitive strain injuries.
150.41 € - 230.02 €
Specifications
Autoclavable | Autoclavable |
---|---|
Number of Channels | 1 |
Sterility | Sterile |
Type | Manual Pipette |
Includes | Lock counter mechanism, disassembly and calibration tool, sample bag with tips, grease |
Product Code | Brand | Volume (Metric) | Increments | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Increments | Price | Quantity & Availability | ||||
10484052
|
Fisherbrand™
FI8-103-00-9 |
5 to 50 μL | 0.50μL |
213.06 €
Each |
In Stock
Log in to see stock. ![]() |
||||
10676813
|
Fisherbrand™
FI8-101-00-9 |
0.5 to 10 μL | 0.01μL |
215.18 €
Each |
In Stock
Log in to see stock. ![]() |
||||
10544073
|
Fisherbrand™
FI8-105-01-9 |
50 to 200 μL | 1.0μL | N/A | |||||
10464412
|
Fisherbrand™
FI8-106-01-9 |
200 to 1000 μL | 5.0μL |
222.60 €
Each |
In Stock
Log in to see stock. ![]() |
||||
10295562
|
Fisherbrand™
FI8-106-00-9 |
100 to 1000 μL | 5.0μL |
150.41 €
Each |
In Stock
Log in to see stock. ![]() |
||||
10091632
|
Fisherbrand™
FI8-104-00-9 |
10 to 100 μL | 0.5μL | N/A | |||||
10063722
|
Fisherbrand™
FI8-102-00-9 |
2 to 20 μL | 0.50μL |
179.69 €
Each |
In Stock
Log in to see stock. ![]() |
||||
10482382
|
Fisherbrand™
FI8-107-00-9 |
500 to 5000 μL | 50μL |
230.02 €
Each |
In Stock
Log in to see stock. ![]() |
||||
Description
- Accurate and reliable pipetting
- Reverse pipetting option for accurate and precise aspirating and dispensing of viscous liquids
- Innovative soft grip prevents transfer of heat to internal components, ensuring accurate pipette operation
- Lock counter mechanism of variable volume pipettes prevents volume changes while pipetting
- Comfortable and effortless pipetting - light weight / ergonomic design for easy operation prevents hand stress. Ergonomic low plunger force minimizes RSI (repetitive strain injury)
- Pipette is fully autoclavable for sterile handling
- Easy and quick maintenance and adjustment with the included disassembly and calibration tool
- No loss of sample - blow out system allows last drop dispensing
- Meets following standards: DIN EN ISO 8655-1 | DIN EN ISO 8655-2 | DIN EN ISO 8655-6 | EU Directive 98/79/EG
Specifications
Autoclavable | |
Sterile | |
Lock counter mechanism, disassembly and calibration tool, sample bag with tips, grease | |
2 Years |
1 | |
Manual Pipette | |
1 Each |