Learn More
Fisherbrand™ Scaffolding Feet
Provides a stable base for laboratory scaffolding. Fisherbrand™ Scaffolding Feet come with holes for screwing the foot to the bench, floor or wall.
Brand: Fisherbrand™ 2117
Description
- Allen key provided
- Socket head screw fixing for rods or tubes of 13mm diameter
- Manufactured from either stainless steel or aluminum
Note:
For use with scaffolding foot adapter (11598112).
Specifications
Aluminum | |
Stand Foot | |
For use with scaffolding foot adapter 11598112. |