Learn More
Thermo Scientific Chemicals Peroxidase, horseradish
Used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody.
1485.00 SEK - 9260.00 SEK
Specifications
Form Lyophilized
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15402879
View Documents Promotion Details
Thermo Scientific Chemicals
J60026.MC
100 mg
1485.00 SEK
100mg
View alternative products
Save Up to 
»
15492869
View Documents Promotion Details
Thermo Scientific Chemicals
J60026.03
1 g
9260.00 SEK
1g
View alternative products
Save Up to 
»
Description
Description
Peroxidase, horseradish is used in the oxidation of luminol to 3-aminophthalate. It is often used in conjugates to determine the presence of a molecular target, especially in the detection of specific protein in a western blot after conjugation with antibody. Further, it is highly reactive toward human tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Lyophilized
Videos
Safety and Handling
Safety and Handling
missing translation for 'einecsNumber' : 232-668-6
Recommended Storage : Store at -20°C