Learn More
Thermo Scientific™ Oxoid™ Penase freeze dried in vial Oxoid
Brand:  Thermo Scientific™ Oxoid™ SR0129B
1498.55 SEK valid until 2025-10-26
Use promo code "24136" to get your promotional price.
Product Code. 10527153
  • 1665.00 SEK / Each
Focus on Agri-Science Testing
Use promo code 24136 at checkout
View Available Promotions
Description
A Bacillus cereus 569/H9 lactamase (E.C.5.2.6) presented as a freeze-dried powder containing buffer salts. Each vial contains a 3,300lU of activity (1 unit of enzyme activity will hydrolyse 1.0μmol of benzylpenicilloic acid per minute pH7.0 and at 25°C).
Used for the inactivation of susceptible beta-lacram antibiotics in preparations, blood or tissue samples.