Learn More
3M™ PELTOR™ Optime™ II Ear Muffs
Developed for demanding, noisy environments and particularly suited for noise levels dominated by low to medium frequencies. 3M™ PELTOR™ Optime™ II Ear Muffs provide ideal hearing protection in environments with substantial industrial noise or construction machinery, like airports and agricultural work.
268.19 SEK - 418.00 SEK
Specifications
Color | Green |
---|---|
Certifications/Compliance | CE Approved |
Plug Shape | Oval |
Material | Stainless Steel |
Description
- Electrically insulated headband (dielectric) version available
- Large space inside cup helps reduce moisture and heat build-up
- Sealing rings filled with a unique combination of liquid and foam
- Easy-to-replace cushions and inserts helps keep them hygienically clean
- Soft, wide cusions help reduce pressure around the ears and improves comfort and wearability
Specifications
Green | |
CE Approved | |
Oval | |
Stainless Steel |