Learn More
Hamilton™ PEEK HPLC Guard Column Starter Kit
4470.00 SEK - 4555.00 SEK
Product Code | Brand | Particle Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Price | Quantity & Availability | |||||
11503334
|
Hamilton™
79831 |
7 μm |
4470.00 SEK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
11523334
|
Hamilton™
79361 |
12 to 20 μm |
4555.00 SEK
Each |
Estimated Shipment: 18-07-2025
Log in to see stock. |
|||||
Specifications
HPLC Guard Column Starter Kit | |
PEEK |