missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDILT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26305-1-AP-20UL
This item is not returnable.
View return policy
Description
Probable redox-inactive chaperone involved in spermatogenesis.Specifications
PDILT | |
Polyclonal | |
Unconjugated | |
PDILT | |
PDIA7, PDILT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
204474 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N807 | |
PDILT | |
PDILT Fusion Protein Ag23675 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction