Learn More
Agilent BioTek Parallel to USB Adapter
For use with Microplate Washers and Readers
Brand: Agilent BioTek 75135
This item is not returnable.
View return policy
Description
- Parallel to USB Adapter
- Use to connect an ELx800, ELx808, FLx800 or uQuant to a USB-only printer
- For use with Agilent BioTek Microplate Washers and Readers
Specifications
Parallel to USB Adapter | |
ELx800, ELx808, FLx800 Fluorescence Reader |