Learn More
Palladium, 5% on activated carbon paste, 5R437, Thermo Scientific™
Product Code. 15486357
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.
Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),
Notes
Sold on a dry weight basis. Unit weight excludes water weight
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]
Specifications
Quantity 10 g
UN Number UN1325
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Chemical Name or Material Palladium, 5% on activated carbon paste, 5R437