missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Aminosalicylic Acid Sodium Salt Dihydrate, approx. 99%, MP Biomedicals™
265.00 SEK - 1240.00 SEK
Chemical Identifiers
CAS | 6018-19-5 |
---|---|
Molecular Formula | C7H10NNaO5 |
Molecular Weight (g/mol) | 211.149 |
InChI Key | GMUQJDAYXZXBOT-UHFFFAOYSA-M |
Synonym | sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium usp |
PubChem CID | 16211148 |
IUPAC Name | sodium;4-amino-2-hydroxybenzoate;dihydrate |
SMILES | C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413191
|
MP Biomedicals
0215036725 |
25 g |
265.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423191
|
MP Biomedicals
0215036790 |
500 g |
1240.00 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
6018-19-5 | |
211.149 | |
sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium usp | |
sodium;4-amino-2-hydroxybenzoate;dihydrate |
C7H10NNaO5 | |
GMUQJDAYXZXBOT-UHFFFAOYSA-M | |
16211148 | |
C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+] |
Specifications
6018-19-5 | |
approx. 99% | |
sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium usp | |
C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+] | |
211.149 | |
211.15 | |
p-Aminosalicylic Acid Sodium Salt Dihydrate |
White | |
C7H10NNaO5 | |
GMUQJDAYXZXBOT-UHFFFAOYSA-M | |
sodium;4-amino-2-hydroxybenzoate;dihydrate | |
16211148 | |
Solid |
Safety and Handling
Recommended Storage : Store at room temperature.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title