Learn More
Thermo Scientific™ Oxoid™ Peptone Bacteriological
The Oxoid Peptone Bacteriological is for use whenever a high quality bacteriological peptone is required.
1750.00 SEK - 27495.00 SEK
Specifications
Content And Storage Ambient & Dry Storage
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Protein Source Beef
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10404013
View Documents Promotion Details
Thermo Scientific™
LP0037B
500 g
1750.00 SEK
500g
Only null left
In Stock
Add to basket
12651555
View Documents
Thermo Scientific™
LP0037K
25 kg
27495.00 SEK
25kg
Description
Description
The Oxoid Peptone Bacteriological is for use whenever a high quality bacteriological peptone is required.
Thermo Scientific™ Oxoid Peptone Bacteriological and its neutralised form are very nutritious all-purpose peptones prepared by the enzymatic digestion of selected animal protein sources. The products are ideal for use in culture media or fermentation applications.
They are specially prepared to be compatible with other refined culture media or fermentation ingredients.
The combination of pancreatin and papain enzyme systems ensures that these bacteriological peptones contain a wide spectrum of polypeptides, reflected in their broad molecular profiles.
The neutralised form evolved from the original to meet those occasions when a slightly higher pH is required. Both, when reconstituted, give a solution free of haze, cloudiness, or precipitation.
With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.
Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.
Supports quality media design:
  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free
Multi-method suitability:
  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture
Specifications
Specifications
Ambient & Dry Storage
Peptone
Beef
Dehydrated Powder
Culture media or fermentation processes
Videos
Safety and Handling
Safety and Handling
missing translation for 'shelfLife' : 1825 Days from Manufacture