Learn More
Thermo Scientific™ Oxoid™ Anaerobic System Schrader Valve Chuck plus Clip, Pressure Gauge
Oxoid Anaerobic System Schrader Valve Chuck plus Clip is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Brand: Thermo Scientific™ HP0020A
1337.89 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Oxoid Anaerobic System Schrader Valve Chuck plus Clip is for use with Thermo Scientific™ Oxoid 3.5L Anaerobic Jar, Part No. HP0011A.
Specifications
Anaerobic System Schrader Valve Chuck plus Clip | |
Pressure Gauge | |
Thermo Scientific Oxoid | |