Learn More
Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel System Replacement Parts
Replace your UVT gel tray, gaskets and external casters with mini gel and multi gel system replacement parts.
205.00 SEK - 925.00 SEK
Specifications
For Use With (Equipment) | Owl™ EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System |
---|---|
Gel Size | Mini |
Product Line | Owl, EasyCast |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11942955
|
Thermo Scientific™
B1A-CST |
Caster |
925.00 SEK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
|||||
11952955
|
Thermo Scientific™
B1A-GK |
Gasket |
205.00 SEK
2 sets |
Estimated Shipment: 13-10-2025
Log in to see stock. |
|||||
11962955
|
Thermo Scientific™
B1A-UVT |
Gel Tray |
925.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
Description
Replace your UVT gel tray, gaskets and external casters with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel and A2-OK Multi Gel System Replacement Parts, for use with EasyCast B1A and A2-OK Multi Gel Mini Gel Systems.
Ideal use with EasyCast B1A and A2-OK Multi Gel Mini Gel Systems.Specifications
Owl™ EasyCast B1A Mini Gel and A2-OK Multi Gel Electrophoresis System | |
Mini | |
Owl, EasyCast |