Learn More
Molecular Probes™ Oregon Green™ 488 Cadaverine, 5-isomer
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
The primary aliphatic amine of the green fluorescent Oregon Green™ cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247). This molecule can also be used as a water-soluble, fixable polar tracer.
Specifications
Specifications
Quantity | 5 mg |
Product Type | Cadaverine |
Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
Chemical Reactivity | Carboxylic Acid, Ketone, Aldehyde |
Shipping Condition | Room Temperature |
Reactive Moiety | Amine, Cadaverine |
Label or Dye | Oregon Green™ 488 |
Label Type | Classic Dyes |
Product Line | Oregon Green™ |