Learn More
Sartorius Stedim Biotech™ Nutrient Pad Sets for E. coli, Coliforms and Enterobacteria
Sartorius Stedim Biotech™ Nutrient Pad Sets for E. coli, Coliforms and Enterobacteria are sterile, dehydrated culture media for microbiological testing.
Brand: Sartorius Stedim Biotech™ 14049--50------N
2700.95 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Practical and easy to handle, they reduce labor and simplify many microbiological testing procedures.
- Nutrient pads are sterile, dehydrated culture media.
- Once they are moistened with 3.0–3.5mL of sterile and demineralized (or distilled) water, they are ready to use immediately.
- Ready to use for up to 24 months.
- Inclusive membranes: supplied with the suitable membrane filters, which are also pre-sterilized and individually packaged.
- Economical.
- Easy to handle.
- Consistent quality.
- Trouble-free storage at room temperature for up to 24 months.
- Compliant with international standards: Aside from the European drinking water directive, they comply with other international regulations and recommendations: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA, etc.
Specifications
Sterile | |
Packaged individually in petri dishes | |
100/Pk. |
Testing for chromogenic coliform (acc. ISO 9308-1) | |
NPS & CN | |
50 mm |