Learn More
Thermo Scientific™ Nunc™ 384-Well, Non-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
3665.00 SEK - 11445.00 SEK
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid With Lid
Culture Area 0.084 cm2/well
Array 16 x 24
View More Specs
Products 3
Product Code Brand No. of Wells No. per Pack No. per Case Price Quantity & Availability  
Product Code Brand No. of Wells No. per Pack No. per Case Price Quantity & Availability  
10162881
View Documents Promotion Details
Thermo Scientific™
242757
384 10 Microplates 30 Microplates
3665.00 SEK
Pack of 30
Only null left
In Stock
Add to basket
10354611
View Documents
Thermo Scientific™
242765
96 1 Microplates 30 Microplates
4410.00 SEK
Pack of 30
Only null left
In Stock
Add to basket
10592835
View Documents Promotion Details
Thermo Scientific™
265202
384 25 Microplates 100 Microplates
11445.00 SEK
Pack of 100
Only null left
Estimated Shipment: 07-07-2025
Add to basket
Description
Description
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Nunc 384-well polystyrene plates have rounded square-well geometry, eliminating wicking.
  • Clear Polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated Polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Customized bar coding is available
  • Flexible working volumes; 10-100μL
Specifications
Specifications
Custom Barcoding Available
With Lid
16 x 24
Clear
Polystyrene
10 to 100 μL
Non-treated
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
0.084 cm2/well
Nunc
Microplate
120 μL
Sterile
Flat Bottom (F-well)