missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSDHL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15111-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked domit disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5′ UTR have been found for this gene.Specifications
NSDHL | |
Polyclonal | |
Unconjugated | |
NSDHL | |
H105E3, NSDHL, Protein H105e3, SDR31E1, XAP104 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18194, 309262, 50814 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15738, Q5PPL3, Q9R1J0 | |
NSDHL | |
NSDHL Fusion Protein Ag7097 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction