missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NR5A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22460-1-AP-150UL
This item is not returnable.
View return policy
Description
NR5A2 (nuclear receptor subfamily 5, group A, member 2), also designated B1-binding factor (B1F or B1F2), CYP7A promoter-binding factor (CPF), fetoprotein-alpha 1 (AFP) transcription factor (FTF, FTZ-F1 or FTZ-F1beta) and liver receptor homolog 1 (LRH-1), is a pre-adipocyte-specific nuclear receptor that regulates expression of aromatase in adipose tissue. NR5A2 belongs to the fushi tarazu factor-1 subfamily of orphan nuclear receptors. NR5A2 transcripts are abundant in the human ovary and testis and are predomitly expressed in tissues of endodermal origin. NR5A2 is a positive transcription factor for ABCG5 and ABCG8 and regulates genes involved in sterol and bile acid secretion from liver and intestine. It induces cell proliferation through the concomitant induction of cyclin D1 and E1, an effect that is potentiated by its interaction with beta-catenin.Specifications
NR5A2 | |
Polyclonal | |
Unconjugated | |
NR5A2 | |
B1 binding factor, B1F, B1F2, CPF, CYP7A promoter binding factor, FTF, FTZ F1, FTZ F1beta, hB1F, hB1F 2, Liver receptor homolog 1, LRH 1, NR5A2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2494, 26424, 60349 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00482, P45448, Q9QWM1 | |
Nr5a2 | |
NR5A2 Fusion Protein Ag18001 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction