Learn More
Bochem™ Nickel Crucibles
1000.00 SEK - 1240.00 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10209170
|
Bochem™
8125 |
50 mL |
1000.00 SEK
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
|||||
10510252
|
Bochem™
8126 |
70 mL |
1030.00 SEK
Each |
Estimated Shipment: 15-07-2025
Log in to see stock. |
|||||
10284601
|
Bochem™
8127 |
130 mL | N/A | ||||||
Specifications
Nickel | |
Crucible |