missing translation for 'onlineSavingsMsg'
Learn More

NFKBIL1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen NFKBIL1
Concentration 0.33 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16864785
View Documents
Proteintech
26273-1-AP-20UL
20 μL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16854785
View Documents
Proteintech
26273-1-AP-150UL
150 μL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NF-kappa-B inhibitor-like protein 1 (NFKBIL1), also named Ikb like and IkbL1, is a 381 amino acid member of the inhibitor of NF-kappaB (IkappaB) family of proteins. NFKBIL1 contains two ANK repeats and is expressed in the rheumatoid synovial lining and sub-lining layers and co-localized in CD68+ and CD3+, but not Factor VIII+ cells. It is reported that defects in NFKBIL1 expression causes of susceptibility to rheumatoid arthritis (RA), which is a systemic inflammatory disease with autoimmune features and affects the joints. Rheumatoid arthritis (RA) is characterized by inflammatory changes in the synovial membranes and articular structures, widespread fibrinoid degeneration of the collagen fibers in mesenchymal tissues, and by atrophy and rarefaction of bony structures.
Specifications

Specifications

NFKBIL1
Western Blot
Unconjugated
Rabbit
Human
Q9UBC1
4795
NFKBIL1 Fusion Protein Ag21978
Primary
-20°C
NFKBIL1
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
I kappa B like protein, IkappaBL, IKBL, LST1, NFKBIL, NFKBIL1
NFKBIL1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.