Learn More
Hamilton™ Neuros™ 7000 Series Modified Microliter Syringes with Knurled Hub Needle
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Brand: Hamilton™ 65459-02
This item is not returnable.
View return policy
Description
- Needle rigidity improves insertion path accuracy
- Minimal tissue damage reduces injection variability
- Reduced sample loss saves money and materials
- Fine gauge needle creates smaller injection sites
- Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Autoclavable | |
30 G | |
0.008 in. | |
0.012 in. | |
0.206 mm | |
0.312 mm | |
0 to 20 mm | |
6000 psig (413.7 bar) | |
115°C | |
7002 KH | |
2 μL |
Tungsten Wire (Plunger) | |
0.008 in. | |
0.20 mm | |
0.308 in. | |
7.82 mm | |
Controlled animal injections | |
Modified Microliter | |
239°F | |
50°F | |
4 | |
Knurled Hub |