Learn More
Thermo Scientific™ Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Ensure contamination-free dispensing with these chemical-resistant wash bottles made with Teflon™ fluoropolymer.
1555.00 SEK - 7805.00 SEK
Specifications
Color | Translucent |
---|---|
Material | FEP |
Closure Included | Yes |
Autoclavable | Autoclavable |
Graduated | No |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10508131
![]() |
Thermo Scientific™
2403-0125 |
125 mL | 24 mm |
1555.00 SEK
Each |
Estimated Shipment: 04-11-2025
Log in to see stock. |
||||
10548701
![]() |
Thermo Scientific™
2403-0250 |
250 mL | 24 mm |
2290.00 SEK
Each |
In Stock
Log in to see stock. |
||||
10170551
![]() |
Thermo Scientific™
2403-0500 |
500 mL | 38-430 mm |
3110.00 SEK
Each |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||
15951491
![]() |
Thermo Scientific™
2403-1000 |
1000 mL | 38-430 mm |
7805.00 SEK
Pack of 2 |
Estimated Shipment: 04-09-2025
Log in to see stock. |
||||
Description
Ensure contamination-free dispensing with Thermo Scientific™ Nalgene™ Wash Bottles made with Teflon™ fluoropolymer, useful for trace element work and other high-purity applications. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymer. Designed to dispense through the top with a closure and stem molded in one piece for reliable service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
- With ETFE screw closure/stem and draw tube
- Demonstrates excellent chemical resistance to nearly any chemical in your lab including solvents—won't become brittle or crack
- Inert fluoropolymer construction is low in extractables and low binding—useful for trace element work
- Tip can be cut back to provide increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
- Autoclavable/Transparent
- Sputter-free, precision stream
Specifications
Translucent | |
Yes | |
No | |
Cylindrical | |
ETFE | |
Yes |
FEP | |
Autoclavable | |
Narrow | |
Wash Bottle | |
Screw Cap |
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.