Learn More
Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles
Prevent separation when squeezing these LDPE economy wash bottles, which feature a tight-fitting draw tube and closure.
285.00 SEK - 1800.00 SEK
Specifications
Color | Translucent |
---|---|
Includes | Stem and draw tube |
Material | LDPE |
Closure Included | Yes |
Graduated | No |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
10709121
![]() |
Thermo Scientific™
2401-0125 |
125 mL | 24 mm |
285.00 SEK
Pack of 6 |
In Stock
Log in to see stock. |
||||
10624301
![]() |
Thermo Scientific™
2401-0250 |
250 mL | 24 mm |
372.00 SEK
Pack of 6 |
In Stock
Log in to see stock. |
||||
11973857
![]() |
Thermo Scientific™
2401-0500 |
500 mL | 28 mm |
1800.00 SEK
Pack of 24 |
In Stock
Log in to see stock. |
||||
10048300
![]() |
Thermo Scientific™
2401-1000 |
1000 mL | 38-430 mm |
347.00 SEK
Pack of 4 |
In Stock
Log in to see stock. |
||||
Description
High quality squeeze bottles at a value price. Thermo Scientific™ Nalgene™ LDPE Economy Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.
Features:
- Polypropylene screw closure/stem with PPCO draw tube
- Tip can be cut back to provide increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
- Flexible LDPE bottle provides soft, ergonomic squeeze
- Sputter-free, precision stream
- Bottle and Carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable LDPE wash bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene LDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene LDPE bottles.
Specifications
Translucent | |
LDPE | |
No | |
Cylindrical | |
Polypropylene | |
Yes |
Stem and draw tube | |
Yes | |
Narrow | |
Economy Wash Bottle | |
Screw Cap |