missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexane, 99%, HPLC grade, Scharlab
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.50 L
Description
- Undergoes highly efficient distillation, solid phase absorption and other treatments to eliminate impurities that could interfere in HPLC analysis
- Microfiltered and packed in amber-coloured bottles (sealed with a PTFE membrane in the cap) to avoid possible contamination
Specifications
Specifications
Melting Point | -94.3°C |
Absorbance | 0.699 AU at 200nm, 0.301 AU at 210nm, 0.046 AU at 230nm |
CAS | 110-54-3 |
Color | Colorless |
Acidity | Max. 0.0002meq/g |
Alkalinity | Max. 0.0002meq/g |
Density | 0.66g/cm3 |
Boiling Point | 69°C |
Quantity | 2.5 L |
Flash Point | −22°C |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction