Learn More
N-cadherin Mouse anti-Human, CoraLite 488, Clone: 1D8B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66219-100UL
Description
The N-cadherin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-cadherin. This antibody recognizes human, pig, mouse, rat antigen. The N-cadherin antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
N-cadherin | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
1000 | |
IgG1 |
Immunofluorescence | |
1D8B3 | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |