missing translation for 'onlineSavingsMsg'
Learn More

n-Butyldimethylchlorosilane, 96%, Thermo Scientific Chemicals

Product Code. 11415577
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 1000-50-6 | C6H15ClSi | 150.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1000-50-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00015727
InChI Key MXOSTENCGSDMRE-UHFFFAOYSA-N
Synonym butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
PubChem CID 66079
SMILES CCCC[Si](C)(C)Cl

Specifications

Boiling Point 138°C
Quantity 100 g
UN Number UN2920
Beilstein 1732573
Flash Point 27°C (80°F)
IUPAC Name butyl-chloro-dimethylsilane
Formula Weight 150.73
Percent Purity 96%
Refractive Index 1.42
Sensitivity Moisture sensitive
Density 0.875
Chemical Name or Material n-Butyldimethylchlorosilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.