Learn More
3M™ Modul-R™ Replacement Clear Lens
Click to view available options
:
Lens Tint: Clear
Description
- Replacement low profile polycarbonate lens
- Clear lens with anti-scratch and anti-fog coating
- For use with the 3M™ Modul-R™ Goggles
Specifications
Specifications
Lens Coating | Scratch-resistant |
Description | Indirect Vented |
Lens Tint | Clear |
Product Type | Replacement lens |
Lens Material | Polycarbonate |