Learn More
3M™ Modul-R™ Half Visor 71361-00003
Click to view available options
:
Lens Tint: Clear
Description
- Half visor with adjustable ventilation system (close or open position)
- Can only be used in conjunction with the 3M Modul-R goggle
Specifications
Specifications
Certifications/Compliance | EN 166: 2001 |
Description | Ventilated Half Visor |
Lens Tint | Clear |
For Use With (Equipment) | Modul-R™ goggles |
For Use With (Application) | Construction |
Lens Material | Polycarbonate |