Learn More
MLKL Mouse anti-Human, CoraLite 488, Clone: 3D4C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66675-100UL
Description
The MLKL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MLKL. This antibody recognizes human, mouse antigen. The MLKL antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
MLKL | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
197259 | |
IgG1 |
Immunofluorescence | |
3D4C6 | |
Mouse | |
Primary | |
Human, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.