Learn More
MLKL Mouse anti-Human, CoraLite 488, Clone: 3D4C6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66675-100UL
Product Code. 16870826
  • 5106.85 SEK / 100µL
Description
The MLKL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MLKL. This antibody recognizes human, mouse antigen. The MLKL antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
MLKL
Monoclonal
CoraLite 488
100 μL
197259
IgG1
Immunofluorescence
3D4C6
Mouse
Primary
Human, Mouse