Learn More
Miele Professional™ Miele Process Documentation
APH 407 Network cable for connection to the network, length 5 m (type 7).
Brand:  Miele Professional™ 7951480
360.06 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15470455
  • 397.00 SEK / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Description
Cable category 6 (color: transparent silver)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Process/Operating Documentation/Communication
Washer-disinfectors, laboratory