Learn More
Ansell Microgard™ 2000 Standard SOCO/CSI Coveralls with Hood
Designed to allow water vapor (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals. Filter 100% of particulates down to 0.01 microns in size.
5630.00 SEK
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Material | Microporous Polyethylene Film |
Color | White |
Disposable | Single-use |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15546524
|
Ansell
WH20B-00128-02 |
Small |
5630.00 SEK
Pack of 40 |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
15556524
|
Ansell
WH20B-00128-03 |
Medium |
5630.00 SEK
Pack of 40 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
15566524
|
Ansell
WH20B-00128-04 |
Large |
5630.00 SEK
Pack of 40 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
15576524
|
Ansell
WH20B-00128-05 |
X-Large |
5630.00 SEK
Pack of 40 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||||
15586524
|
Ansell
WH20B-00128-06 |
2X-Large |
5630.00 SEK
Pack of 40 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
15596524
|
Ansell
WH20B-00128-07 |
3X-Large |
5630.00 SEK
Pack of 40 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
|||||
Description
- Self adhesive pockets can be positioned anywhere on the garment to secure equipment
- Finger loops prevent sleeve movement when working above your head
- Protection from biological agents in the highest performance class according to EN14126
- Anti-static according to EN 1149-5
- Elasticated hood, wrists, waist and ankles
- 2-way front zipper with resealable storm flap
- 2-piece hood with chinstrap
- Dual finger loops
- Includes 2 pockets
Specifications
General Purpose Coveralls | |
Microporous Polyethylene Film | |
Single-use | |
Elastic | |
Hooded | |
Front Zipper with Storm Flap | |
Non-sterile | |
Hood with chinstrap, dual finger loops | |
2 |
Unisex | |
White | |
Elastic | |
Elastic | |
2-panel with Chin Strap | |
Bound | |
Cat III Type 5-B, 6-B, EN1073-2, EN14126, EN1149-1, DIN 32781 | |
WH20-B-00-128 | |
Includes Hood with Chinstrap, Dual Fingerloops |