Learn More
MICA/MICB Mouse anti-Human, APC, Clone: 6D4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech APC-65161-100TESTS
Description
The MICA/MICB antibody from Proteintech is a mouse monoclonal antibody generated with transfected c1r cells expressing mica of human MICA/MICB. This antibody recognizes human antigen. The MICA/MICB antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
MICA/MICB | |
Monoclonal | |
0.1 mg/mL | |
Mouse | |
Primary | |
Human |
Flow Cytometry | |
6D4 | |
APC | |
100 μg | |
4276 | |
IgG2a |