Learn More
Mettler Toledo™ Glass Cylinder For Titration Sensor Interchangeable Burette
Replacement part for DV1005 or DV1020 titration burettes.
Brand: Mettler Toledo™ 51107102
This item is not returnable.
View return policy
Description
Replacement part for DV1005 or DV1020 titration burettes.Compliance
ISO 8655-3
Specifications
Cylinder | |
Glass | |
20 mL Burettes |