Learn More
IKA™ MATRIX Cover
Cover for MATRIX Orbital Delta F1.5/ FP/ F0.5/ F2.0, MATRIX PCR 96 / PCR 384 attachments, MATRIX 0.5 - 2.0 ml as well as plate attachments.
Brand:  IKA™ 0030001253
Product Code. 16248701
  • 1339.13 SEK / Each
Only null left
Estimated Shipment: 06-08-2025
Description
For even tempering and to protect the samples from unwanted exposure to light.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cover
147 x 104 x 45 mm
Even tempering and to protect the samples from unwanted exposure to light.
Cover for MATRIX Orbital Delta F1.5/FP/F0.5/F2.0, MATRIX PCR 96 /PCR 384 Attachments, MATRIX 0.5 - 2.0 mL as Well as IKA CARRIER.