Learn More
Masterflex™ L/S™ Small Cartridges for Multichannel Cartridge Pump Heads
Snap these cartridges in and out for fast tubing changes. Use Masterflex™ L/S™ Small Cartridges with L/S™ multichannel cartridge pump heads for synchronous, multichannel pumping.
Brand: Masterflex™ 07519-80
1953.62 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Snap these cartridges in and out for fast tubing changes. Use Masterflex™ L/S™ Small Cartridges with L/S™ multichannel cartridge pump heads for synchronous, multichannel pumping.
- No need to remove pump head from the drive
- Load pump heads with 1 to 8 cartridges
- Mix large and small cartridges
- One large cartridge occupies the same space as two small ones
- Cartridge heads are available with three or four rollers; the greater the number of rollers, the lower the pulsation in the flow
- Require drives with a minimum of 1⁄10hp and a maximum of 600rpm
Specifications
For synchronous, multichannel pumping | |
Multiple | |
1 Each |
Microbore and Precision Tubing | |
Adjustable | |
Cartridge Pump Head |