Learn More
Magnesium Sulfate, 7-Hydrate, Crystal, Multi-Compendial, U.S.P., J.T. Baker™
Product Code. p-5390786
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.50kg
500g
Chemical Identifiers
CAS 10034-99-8
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Specifications
Physical Form Crystals
Quantity 500 g
Formula Weight 246.47
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.67kg
Chemical Name or Material Magnesium Sulfate, 7-Hydrate