Learn More
Cole Parmer Luer Fittings
Cole Parmer Luer Fittings feature exceptional durability and are easy to disassemble for cleaning
Brand:  Cole Parmer 41507-26
Product Code. 15162359
  • 1480.00 SEK / Each
Only null left
Estimated Shipment: 16-09-2025
Description
Cole Parmer Luer Fittings feature exceptional durability and are easy to disassemble for cleaning
Ideal for harsh chemical environments
Specifications
Specifications
Male luer lock x 1/16 in. hose barb
Stainless Steel
15.748 mm
5/8 in.
Autoclavable
5/8 in.
1 Ea.
Luer fittings, male luer lock x 1/16 in. hose barb