Learn More
Lithium Hydroxide Monohydrate, Honeywell Fluka™
Product Code. 15631240 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 1 kg
100 kg
Purum,≥98.5%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1310-66-3
Molecular Formula H3LiO2
Molecular Weight (g/mol) 41.96
MDL Number MFCD00149772
InChI Key GLXDVVHUTZTUQK-UHFFFAOYSA-M
Synonym lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water
PubChem CID 168937
SMILES [Li+].O.[OH-]
Specifications
Quantity 6 x 1 kg
Linear Formula LiOH · H2O
UN Number UN2680
IUPAC Name lithium(1+) hydrate hydroxide
Formula Weight 41.96
Percent Purity ≥98.5%
Grade purum
Chemical Name or Material Lithium Hydroxide Monohydrate