Learn More
Fisherbrand™ Leaf Calibration Weight Set, Class F1
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Leaf Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
Brand:  Fisherbrand™ FB67825
3450.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15545109
  • 4870.00 SEK / 1 unit
Only null left
Estimated Shipment: 09-09-2025
Add to basket
Description
Description
  • Class F1 calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
F1
1 to 500 mg
1 mg to 500 mg