Learn More
UVP Lamp 3Uv-36 3Uv Multi-Wavelength Supplied with 1 X 254Nm
Brand:  UVP 95-0342-02
Product Code. 12883898
  • 19580.00 SEK / Each
Only null left
Estimated Shipment: 22-09-2025
Description
1 x 302nm and 1 x 365nm tube 230V 50Hz 6W UVP