Learn More
Labconco™ Stainless-Steel Racks for RapidVap™ Vacuum and N2 Evaporation Systems
Stainless steel racks hold RapidVap tubes upright.
3255.00 SEK
Specifications
For Use With (Equipment) | RapidVap vacuum and N2 evaporation system |
---|---|
Material | Stainless Steel |
No. of Places | 8 |
Type | Tube Rack |
Weight (English) | 2 lb. |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11731559
|
Labconco™
7486300 |
170 mL |
3255.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11741559
|
Labconco™
7487600 |
600 mL |
3255.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Stainless steel racks hold RapidVap tubes upright.
Stainless steel racks have eight places to hold RapidVap tubes upright. For use with 600 ml tubes.Specifications
RapidVap vacuum and N2 evaporation system | |
8 | |
2 lb. |
Stainless Steel | |
Tube Rack | |
0.9 kg |