Learn More
Labconco™ Flask Holder
Helps to prevent sample melt back
835.00 SEK - 915.00 SEK
Specifications
Type | Flask Holder |
---|---|
No. of Shelves | 0 |
Material | Stainless Steel |
For Use With (Application) | To prevent sample melt back |
For Use With (Equipment) | For fast freeze™ flasks, inner flasks 7542000 (40ml) and 7542200 (80ml) and outer flasks 75411 |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16895883
|
Labconco™
7543300 |
Small |
835.00 SEK
Each |
Estimated Shipment: 30-09-2025
Log in to see stock. |
|||||
16805893
|
Labconco™
7543400 |
Large |
915.00 SEK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
Description
- Made of stainless-steel
- Allows a small fast-freeze flask bottom containing the sample to be placed inside a larger fast-freeze flask
Specifications
Flask Holder | |
Stainless Steel | |
For fast freeze™ flasks, inner flasks 7542000 (40ml) and 7542200 (80ml) and outer flasks 75411 | |
5 in. | |
1 lb. |
0 | |
To prevent sample melt back | |
3.45 in. | |
12.7 cm | |
0.5 kg |