Learn More
Labconco™ Drying Train: Oxygen and Moisture Tubing Kit
Accessories for Labconco Drying Train
Brand:  Labconco™ 5242500
5377.98 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15548723
  • 5940.00 SEK / Each
Includes: Tubing kits are composed of 25 ft. fluorinated ethylene propylene (FEP) tubing and eight tube inserts
Description
Description
For all glove boxes
Specifications
Specifications
3 lb.
25 ft. FEP Tubing and Eight Tube Inserts to Connect Drying Train Components
All Glove Box
1.4 Kg
Enclosure Kit