missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, 99+%, Monosodium Salt, MP Biomedicals

Product Code. p-2625346 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 142-47-2
Molecular Formula C5H8NNaO4
Molecular Weight (g/mol) 169.11
MDL Number MFCD00150138
InChI Key LPUQAYUQRXPFSQ-UHFFFAOYNA-M
Synonym natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt
PubChem CID 86748263
IUPAC Name sodium 2-amino-4-carboxybutanoate
SMILES [Na+].NC(CCC(O)=O)C([O-])=O

Specifications

Quantity 100 g
Formula Weight 169.1
Grade Cell Culture
Chemical Name or Material L-Glutamic Acid, 99+%, Monosodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.