Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Rotary Evaporator Glass Vial Connecting Adapters
Ideal for use with small samples
Brand: DWK Life Sciences 747130-1314
Description
Ideal for use with small samples
Connect standard rotary evaporator traps to vials with variety of GPI thread sizes. Typical receivers include Accuform™ 1 to 10mL vials (06-100C, 06-100E, 06-100F, 06-100G). Any standard vial with GPI threads can be used.
Specifications
Glass | |
Standard Taper Joint: 14/20 | |
Vial Connecting Adapter | |
13-425 |