Learn More
KIAA0368 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25684-1-AP-150UL
 View more versions of this product
Product Code. 16894245
  • 4652.90 SEK / 150µL
Description
Ecm29 appears to be one of three proteins which associate significantly with proteasomes. Ecm29 has been shown to enhance the stability of the proteasome. Ecm29 deletion has been shown to produce 26S proteasomes which dissociated in the absence of ATP. It was thus suggested that Ecm29 tethers the regulatory complex to the proteasome.
Specifications
Specifications
KIAA0368
Polyclonal
Unconjugated
ECPAS
ECM29, extracellular mutant 29, KIAA0368, KIAA1962, RP11 386D8.2
Rabbit
Antigen Affinity Chromatography
RUO
23392
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5VYK3
ECPAS
KIAA0368 Fusion Protein Ag22309
150 μL