Learn More
Kimtech™ JACKSON SAFETY™ NRD Dual Valved Respirators
Providing filtration for harmful substances to help you breathe freely and get the job done safely. Kimberly-Clark™ JACKSON SAFETY™ NRD Dual Valved Respirators are latex-free, moulded and single-use.
Specifications
Certifications/Compliance | EN149:2001+A1:2009 |
---|---|
Disposable | Single-use |
Product Type | Respiratory Safety Mask |
For Use With (Application) | For filtering Metal fumes, fine dust, water and oil-based mist |
Includes | Valved adjustable nose clips |
Description
- Available in two different performance levels
- Shapes easily to face, keeping it cooler and drier
- Ideal for filtering fine dust, water- and oil-based mist, metal fumes and pollution
- With soft foam nose chamber, convex shape and extra-wide adjustable head straps
Specifications
EN149:2001+A1:2009 | |
Respiratory Safety Mask | |
Valved adjustable nose clips | |
10/Pk. |
Single-use | |
For filtering Metal fumes, fine dust, water and oil-based mist | |
Yes |