missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl Acetate, 99%, Honeywell™

Product Code. 15605780 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
25 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-19-0
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008932
InChI Key GJRQTCIYDGXPES-UHFFFAOYSA-N
Synonym isobutyl acetate, isobutyl ethanoate, acetic acid, 2-methylpropyl ester, 2-methyl-1-propyl acetate, 2-methylpropyl ethanoate, acetic acid, isobutyl ester, acetic acid isobutyl ester, acetate d'isobutyle, iso-butyl acetate, beta-methylpropyl ethanoate
PubChem CID 8038
ChEBI CHEBI:50569
IUPAC Name 2-methylpropyl acetate
SMILES CC(C)COC(C)=O

Specifications

Melting Point -99°C
Color Colorless
Density 0.867g/mL at 25°C
Boiling Point 117°C
Explosion Limit 0.105
Flash Point 22°C (71.6°F)
Ignition Point 793°F
Packaging Glass Bottle
Linear Formula CH3COOCH2CH(CH3)2
Refractive Index n20/D 1.39
Quantity 1 L
UN Number UN1213
Beilstein 1741909
Vapor Density >4 (vs air)
Formula Weight 116.16g/mol
Vapor Pressure 15mmHg (20°C)
Percent Purity 99%
Chemical Name or Material Isobutyl acetate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.